Ethyl 2-(3-(aminomethyl)oxetan-3-yl)acetate hemioxalate
Lieferant: Apollo Scientific
|
Gefahr
|
OR55205-1G
APOSOR55205-1GEA
945
EUR
APOSOR55205-1G
Ethyl 2-(3-(aminomethyl)oxetan-3-yl)acetate hemioxalate
Ethyl 2-(3-(aminomethyl)oxetan-3-yl)acetate hemioxalate
Formel:
2C₈H₁₅NO₃·C₂H₂O₄ Molecular Weight: 436,454 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16652331 CAS-Nummer: 1242267-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...