4-Bromo-1H-pyrrolo[2,3-c]pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-6-azaindole

OR55212-1G
APOSOR55212-1GEA 37.7 EUR
APOSOR55212-1G
4-Bromo-1H-pyrrolo[2,3-c]pyridine 97%
4-Bromo-1H-pyrrolo[2,3-c]pyridine
Formel: C₇H₅BrN₂
Molecular Weight: 197,04 g/mol
Schmelzpunkt: 188…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847768
CAS-Nummer: 69872-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR