4-Bromo-2,5-dimethylbenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR55236-500MG
APOSOR55236-500MGEA 222 EUR
APOSOR55236-500MG
4-Bromo-2,5-dimethylbenzaldehyde
4-Bromo-2,5-dimethylbenzaldehyde
Formel: C₉H₉BrO
Molecular Weight: 213,071 g/mol
Schmelzpunkt: 58…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16295236
CAS-Nummer: 88111-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR