4-Methoxydiphenylamin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methoxy-N-phenylaniline
OR55248-25G
APOSOR55248-25GEA
82.8
EUR
APOSOR55248-25G
4-Methoxydiphenylamin 97%
4-Methoxydiphenylamin
Formel:
C₁₃H₁₃NO Molecular Weight: 199,2500 g/mol Siedepunkt: 195 °C/12mmHg Schmelzpunkt: 106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00228649 CAS-Nummer: 1208-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...