2-Iodo-1,4-benzoquinone

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Iodocyclohexa-2,5-diene-1,4-dione

OR55251-1G OR55251-250MG
APOSOR55251-1GEA 393 EUR
APOSOR55251-1G APOSOR55251-250MG
2-Iodo-1,4-benzoquinone
2-Iodo-1,4-benzoquinone
Formel: C₆H₃IO₂
Molecular Weight: 233.991 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01663089
CAS-Nummer: 3958-83-6
UN: 2587
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR