2-Iodo-1,4-benzoquinone
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Iodocyclohexa-2,5-diene-1,4-dione
OR55251-1G
OR55251-250MG
APOSOR55251-1GEA
393
EUR
APOSOR55251-1G
APOSOR55251-250MG
2-Iodo-1,4-benzoquinone
2-Iodo-1,4-benzoquinone
Formel:
C₆H₃IO₂ Molecular Weight: 233.991 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01663089 CAS-Nummer: 3958-83-6 UN: 2587 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...