2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Chloroethyl)isoindoline-1,3-dione
OR55253-1G
APOSOR55253-1GEA
74.1
EUR
APOSOR55253-1G
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione
2-(2-Chloroethyl)-1H-isoindole-1,3(2H)-dione
Formel:
C₁₀H₈ClNO₂ Molecular Weight: 209,63 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129356 CAS-Nummer: 6270-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...