Pent-4-en-2-amine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR55255-250MG
APOSOR55255-250MGEA 464 EUR
APOSOR55255-250MG
Pent-4-en-2-amine hydrochloride 95%
Pent-4-en-2-amine hydrochloride
Formel: C₅H₁₁N·ClH
Molecular Weight: 121,608 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19382008
CAS-Nummer: 1315366-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR