trans-2-Amino-1-hydroxyindane hydrochloride

Lieferant: Apollo Scientific
Achtung

OR55258-1G
APOSOR55258-1GEA 2030 EUR
APOSOR55258-1G
trans-2-Amino-1-hydroxyindane hydrochloride
trans-2-Amino-1-hydroxyindane hydrochloride
Formel: C₉H₁₁NO·ClH
Molecular Weight: 185,651 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026661
CAS-Nummer: 13575-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR