2-Chloro-4-(methylthio)pyridine

Lieferant: Apollo Scientific
Achtung

OR55260-250MG OR55260-1G
APOSOR55260-250MGEA 174 EUR
APOSOR55260-250MG APOSOR55260-1G
2-Chloro-4-(methylthio)pyridine
2-Chloro-4-(methylthio)pyridine
Formel: C₆H₆ClNS
Molecular Weight: 159.637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07437890
CAS-Nummer: 71506-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR