5-{[(Benzyloxy)carbonyl]amino}pentansäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Benzyloxycarbonylamino)valeric acid, 5-(Carbobenzoxyamino)pentanoic Acid, N-Cbz-5-aminovaleric Acid, 5-(((Benzyloxy)carbonyl)amino)pentanoic acid, N-Cbz-5-aminopentanoic Acid

OR55278-25G
APOSOR55278-25GEA 116 EUR
APOSOR55278-25G
5-{[(Benzyloxy)carbonyl]amino}pentansäure
5-{[(Benzyloxy)carbonyl]amino}pentansäure
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251.28 g/mol
Schmelzpunkt: 99…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00188458
CAS-Nummer: 23135-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR