1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-(Azetidin-1-yl)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
OR55283-250MG
APOSOR55283-250MGEA
459
EUR
APOSOR55283-250MG
1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-[2-(Azetidin-1-yl)ethyl]-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel:
C₁₄H₂₄BN₃O₂ Molecular Weight: 277,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27987628 CAS-Nummer: 1422126-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...