1-(1-Boc-piperidino)-4-brom-1H-pyrazol

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(4-bromo-1H-pyrazol-1-yl)piperidine-1-carboxylate, 1-(4-Boc-1-piperidinyl)-4-bromopyrazole

OR55287-25G OR55287-5G
APOSOR55287-25GEA 203 EUR
APOSOR55287-25G APOSOR55287-5G
1-(1-Boc-piperidino)-4-brom-1H-pyrazol
1-(1-Boc-piperidino)-4-brom-1H-pyrazol
Formel: C₁₃H₂₀BrN₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09831980
CAS-Nummer: 877399-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR