1-Bromo-4-nitronaphthalene
Lieferant: Apollo Scientific
Synonyme:
1Bromo-4-nitronaphthalene
OR55291-1G
APOSOR55291-1GEA
77
EUR
APOSOR55291-1G
1-Bromo-4-nitronaphthalene
1-Bromo-4-nitronaphthalene
Formel:
C₁₀H₆BrNO₂ Molecular Weight: 252,064 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00971828 CAS-Nummer: 4236-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...