5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine

OR55297-1G
APOSOR55297-1GEA 720 EUR
APOSOR55297-1G
5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine
5,7-Dimethyl-1,2,3,4-tetrahydro-1,8-naphthyridine
Formel: C₁₀H₁₄N₂
Molecular Weight: 162.232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13705330
CAS-Nummer: 65541-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR