2,3-Dimethoxy-2,3-dimethylbutane

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dimethoxy-2,3-dimethylbutane

OR55306-1G
APOSOR55306-1GEA 405 EUR
APOSOR55306-1G
2,3-Dimethoxy-2,3-dimethylbutane
2,3-Dimethoxy-2,3-dimethylbutane
Formel: C₈H₁₈O₂
Molecular Weight: 146,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28137218
CAS-Nummer: 6091-59-4
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR