tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR55309-1G
APOSOR55309-1GEA 66.7 EUR
APOSOR55309-1G
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate 97%
tert-Butyl 3,6-diazabicyclo[3.1.1]heptane-6-carboxylate
Formel: C₁₀H₁₈N₂O₂
Molecular Weight: 198,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17016736
CAS-Nummer: 869494-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR