4-Chlor-(2-ethoxycarbonyl)phenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-2-ethoxycarbonylphenylboronic acid

OR5531-1G
APOSOR5531-1GEA 297 EUR
APOSOR5531-1G
4-Chlor-(2-ethoxycarbonyl)phenylboronsäure 95%
4-Chlor-(2-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228,4400 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659823
CAS-Nummer: 850568-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR