2,4-Dichloro-5-cyclopropylpyrimidine 95%

Lieferant: Apollo Scientific

OR55311-1G
APOSOR55311-1GEA 364 EUR
APOSOR55311-1G
2,4-Dichloro-5-cyclopropylpyrimidine 95%
2,4-Dichloro-5-cyclopropylpyrimidine
Formel: C₇H₆Cl₂N₂
Molecular Weight: 189,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18533532
CAS-Nummer: 1190379-86-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR