2-(4-Methylbenzoyl)pyridine 95%

Lieferant: Apollo Scientific

OR55312-250MG
APOSOR55312-250MGEA 394 EUR
APOSOR55312-250MG
2-(4-Methylbenzoyl)pyridine 95%
2-(4-Methylbenzoyl)pyridine
Formel: C₁₃H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD07699227
CAS-Nummer: 78539-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR