1-Bromo-2-methoxy-4-methyl-5-nitrobenzene 97%
Lieferant: Apollo Scientific
OR55313-1G
APOSOR55313-1GEA
350
EUR
APOSOR55313-1G
1-Bromo-2-methoxy-4-methyl-5-nitrobenzene 97%
1-Bromo-2-methoxy-4-methyl-5-nitrobenzene
Formel:
C₈H₈BrNO₃ Molecular Weight: 246,058 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007872 CAS-Nummer: 861076-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...