3-Amino-5-(methoxycarbonyl)benzeneboronic acid hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Amino-5-(methoxycarbonyl)phenyl)boronic acid hydrochloride

OR5533-1G
APOSOR5533-1GEA 297 EUR
APOSOR5533-1G
3-Amino-5-(methoxycarbonyl)benzeneboronic acid hydrochloride 97%
3-Amino-5-(methoxycarbonyl)benzeneboronic acid hydrochloride
Formel: C₈H₁₀BNO₄·HCl
Molecular Weight: 231,44 g/mol
Schmelzpunkt: 220…226 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04971992
CAS-Nummer: 380430-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR