N-(3-(4,5-Dihydrothiazol-2-yl)thiazolidin-2-ylidene)-5H-dibenzo[a,d][7]annulen-5-amine 97%

Lieferant: Apollo Scientific
Achtung

OR55331-10MG
APOSOR55331-10MGEA 21.9 EUR
APOSOR55331-10MG
N-(3-(4,5-Dihydrothiazol-2-yl)thiazolidin-2-ylidene)-5H-dibenzo[a,d][7]annulen-5-amine 97%
N-(3-(4,5-Dihydrothiazol-2-yl)thiazolidin-2-ylidene)-5H-dibenzo[a,d][7]annulen-5-amine
Formel: C₂₁H₁₉N₃S₂
Molecular Weight: 377,526 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17676143
CAS-Nummer: 1072145-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR