Isopropyl 5-chloro-4-methyl-2-nitrobenzoate
Lieferant: Apollo Scientific
Synonyme:
Propan-2-yl 5-chloro-4-methyl-2-nitrobenzoate
OR55336-1G
APOSOR55336-1GEA
2050
EUR
APOSOR55336-1G
Isopropyl 5-chloro-4-methyl-2-nitrobenzoate
Isopropyl 5-chloro-4-methyl-2-nitrobenzoate
Formel:
C₁₁H₁₂ClNO₄ Molecular Weight: 257,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27976845 CAS-Nummer: 1204518-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...