1-Methyl-1H-pyrazol-5-ol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-pyrazol-5-ol, 5-Hydroxy-1-methyl-1H-pyrazole
OR55337-1G
APOSOR55337-1GEA
21.9
EUR
APOSOR55337-1G
1-Methyl-1H-pyrazol-5-ol 97%
1-Methyl-1H-pyrazol-5-ol
Formel:
C₄H₆N₂O Molecular Weight: 98,1032 g/mol Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864418 CAS-Nummer: 33641-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...