2-(Boc-amino)-4-[(Boc-amino)methyl]oxazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl N-[4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]-1,3-oxazol-2-yl]carbamate
OR55338-1G
APOSOR55338-1GEA
655
EUR
APOSOR55338-1G
2-(Boc-amino)-4-[(Boc-amino)methyl]oxazole 95%
2-(Boc-amino)-4-[(Boc-amino)methyl]oxazole
Formel:
C₁₄H₂₃N₃O₅ Molecular Weight: 313.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31536689 CAS-Nummer: 2135339-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...