Zn(II) meso-Tetra(4-sulphonatophenyl)porphine

Lieferant: Apollo Scientific
Achtung

Synonyme: Meso-tetra(4-sulphonatophenyl)porphine Zn(II), ZnTPPS

OR55341-100MG
APOSOR55341-100MGEA 87 EUR
APOSOR55341-100MG
Zn(II) meso-Tetra(4-sulphonatophenyl)porphine
Zn(II) meso-Tetra(4-sulphonatophenyl)porphine
Formel: C₄₄H₂₈N₄O₁₂S₄Zn
Molecular Weight: 998,382 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31619566
CAS-Nummer: 56047-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR