4-Isobutyl-3-nitrophenylboronsäure

Lieferant: Apollo Scientific

OR5535-1G
APOSOR5535-1GEA 271 EUR
APOSOR5535-1G
4-Isobutyl-3-nitrophenylboronsäure
4-Isobutyl-3-nitrophenylboronsäure
Formel: C₁₀H₁₄BNO₄
MDL: MFCD04115653
CAS-Nummer: 850568-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR