3-(Dimethoxymethyl)aniline 95%

Lieferant: Apollo Scientific

OR55352-5G
APOSOR55352-5GEA 1180 EUR
APOSOR55352-5G
3-(Dimethoxymethyl)aniline 95%
3-(Dimethoxymethyl)aniline
Formel: C₉H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03643881
CAS-Nummer: 53663-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR