4-(Dimethoxymethyl)aniline 95%

Lieferant: Apollo Scientific

OR55356-5G
APOSOR55356-5GEA 1180 EUR
APOSOR55356-5G
4-(Dimethoxymethyl)aniline 95%
4-(Dimethoxymethyl)aniline
Formel: C₉H₁₃NO₂
Molecular Weight: 167,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20542810
CAS-Nummer: 111616-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR