2-(Dimethoxymethyl)-4-methoxyaniline 95%
Lieferant: Apollo Scientific
OR55361-5G
APOSOR55361-5GEA
1180
EUR
APOSOR55361-5G
2-(Dimethoxymethyl)-4-methoxyaniline 95%
2-(Dimethoxymethyl)-4-methoxyaniline
Formel:
C₁₀H₁₅NO₃ Molecular Weight: 197,231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31614275 CAS-Nummer: 2244085-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...