4-Bromo-2-(dimethoxymethyl)aniline 95%
Lieferant: Apollo Scientific
OR55363-5G
APOSOR55363-5GEA
1180
EUR
APOSOR55363-5G
4-Bromo-2-(dimethoxymethyl)aniline 95%
4-Bromo-2-(dimethoxymethyl)aniline
Formel:
C₉H₁₂BrNO₂ Molecular Weight: 246,101 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31614276 CAS-Nummer: 2244086-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...