3-Hydroxy-2-methoxy-6-(dimethoxymethyl)aniline 95%
Lieferant: Apollo Scientific
OR55366-5G
APOSOR55366-5GEA
1280
EUR
APOSOR55366-5G
3-Hydroxy-2-methoxy-6-(dimethoxymethyl)aniline 95%
3-Hydroxy-2-methoxy-6-(dimethoxymethyl)aniline
Formel:
C₁₀H₁₅NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31614278 CAS-Nummer: 2244083-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...