4-Chloro-3-(dimethoxymethyl)aniline 95%

Lieferant: Apollo Scientific

OR55369-5G
APOSOR55369-5GEA 1180 EUR
APOSOR55369-5G
4-Chloro-3-(dimethoxymethyl)aniline 95%
4-Chloro-3-(dimethoxymethyl)aniline
Formel: C₉H₁₂ClNO₂
Molecular Weight: 201,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31614279
CAS-Nummer: 2244085-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR