5-Acetamido-2-nitrophenylboronsäure 96%
Lieferant: Apollo Scientific
Synonyme:
(5-Acetamido-2-nitro)benzeneboronicacid
OR5539-1G
OR5539-5G
OR5539-10G
APOSOR5539-1GEA
143
EUR
APOSOR5539-1G
APOSOR5539-5G
APOSOR5539-10G
5-Acetamido-2-nitrophenylboronsäure 96%
5-Acetamido-2-nitrophenylboronsäure
Formel:
C₈H₉BN₂O₅ Schmelzpunkt: 193…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659829 CAS-Nummer: 78887-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...