2'-Aminoacetophenon 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
o-Aminoacetophenon
OR55399-100G
OR55399-1KG
OR55399-500G
APOSOR55399-100GEA
39.3
EUR
APOSOR55399-100G
APOSOR55399-1KG
APOSOR55399-500G
2'-Aminoacetophenon 95%
2'-Aminoacetophenon
Formel:
C₈H₉NO Molecular Weight: 135.17 g/mol Siedepunkt: 252 Schmelzpunkt: 20 Lagertemperatur: Kühlschrank |
MDL:
MFCD00007717 CAS-Nummer: 551-93-9 Merck Index: 13,00412 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...