2'-Aminoacetophenon 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: o-Aminoacetophenon

OR55399-100G OR55399-1KG OR55399-500G
APOSOR55399-100GEA 39.3 EUR
APOSOR55399-100G APOSOR55399-1KG APOSOR55399-500G
2'-Aminoacetophenon 95%
2'-Aminoacetophenon
Formel: C₈H₉NO
Lagertemperatur: Kühlschrank
MDL: MFCD00007717
CAS-Nummer: 551-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR