5-Bromo-6-methoxy-1-indanone 97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-6-methoxy-2,3-dihydroinden-1-one, 5-Bromo-6-methoxy-2,3-dihydro-1H-inden-1-one

OR55405-1G
APOSOR55405-1GEA 91.9 EUR
APOSOR55405-1G
5-Bromo-6-methoxy-1-indanone 97%
5-Bromo-6-methoxy-1-indanone
Formel: C₁₀H₉BrO₂
Schmelzpunkt: 150…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11044062
CAS-Nummer: 187872-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR