Thiomorpholine, N-BOC protected

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl thiomorpholine-4-carboxylate

OR55408-250MG OR55408-1G OR55408-5G
APOSOR55408-250MGEA 26.1 EUR
APOSOR55408-250MG APOSOR55408-1G APOSOR55408-5G
Thiomorpholine, N-BOC protected
Thiomorpholine, N-BOC protected
Formel: C₉H₁₇NO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD14635788
CAS-Nummer: 220655-09-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR