4-Benzyloxy-3-methylphenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Benzyloxy)-3-methylphenyl)boronic acid, 4-Benzyloxy-3-methylbenzeneboronic acid

OR5541-5G
APOSOR5541-5GEA 87 EUR
APOSOR5541-5G
4-Benzyloxy-3-methylphenylboronsäure 96%
4-Benzyloxy-3-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242,08 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06409944
CAS-Nummer: 338454-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR