5'-Bromo-2'-Methoxy-3'-methylacetophenone 95+%

Lieferant: Apollo Scientific

OR55444-1G
APOSOR55444-1GEA 873 EUR
APOSOR55444-1G
5'-Bromo-2'-Methoxy-3'-methylacetophenone 95+%
5'-Bromo-2'-Methoxy-3'-methylacetophenone
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243.097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621110
CAS-Nummer: 1781561-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR