5-Bromo-2-Methoxy-3-methylphenacyl bromide 95+%

Lieferant: Apollo Scientific

OR55447-1G
APOSOR55447-1GEA 873 EUR
APOSOR55447-1G
5-Bromo-2-Methoxy-3-methylphenacyl bromide 95+%
5-Bromo-2-Methoxy-3-methylphenacyl bromide
Formel: C₁₀H₁₀Br₂O₂
Molecular Weight: 321,993 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31621111
CAS-Nummer: 2244084-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR