ɑ-Methyl-γ-butyrolactone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyldihydrofuran-2(3H)-one, Dihydro-3-methyl-2(3H)-furanone, α-Methyl-γ-butyrolactone

OR55456-25G
APOSOR55456-25GEA 129 EUR
APOSOR55456-25G
ɑ-Methyl-γ-butyrolactone 98%
alpha-Methyl-gamma-butyrolactone
Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005396
CAS-Nummer: 1679-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR