Imidazo[1,2-a]pyridin-6-ylmethanamine hydrochloride 96%
Lieferant: Apollo Scientific
|
Gefahr
|
OR55457-1G
APOSOR55457-1GEA
293
EUR
APOSOR55457-1G
Imidazo[1,2-a]pyridin-6-ylmethanamine hydrochloride 96%
Imidazo[1,2-a]pyridin-6-ylmethanamine hydrochloride
Formel:
C₈H₉N₃·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199946 CAS-Nummer: 1352305-21-5 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...