1-Boc-5-methyl-2-indolboronsäure

Lieferant: Apollo Scientific

Synonyme: (1-(tert-Butoxycarbonyl)-5-methyl-1H-indol-2-yl)boronic acid, 5-Methyl-1H-indole-2-boronic acid, N-BOC protected

OR5546-1G
APOSOR5546-1GEA 230 EUR
APOSOR5546-1G
1-Boc-5-methyl-2-indolboronsäure
1-Boc-5-methyl-2-indolboronsäure
Formel: C₁₄H₁₈BNO₄
Molecular Weight: 275,11 g/mol
MDL: MFCD04114580
CAS-Nummer: 475102-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR