Methyl 2-chloro-5-(methylamino)benzoate 95%

Lieferant: Apollo Scientific

OR55472-1G OR55472-500MG
APOSOR55472-1GEA 510 EUR
APOSOR55472-1G APOSOR55472-500MG
Methyl 2-chloro-5-(methylamino)benzoate 95%
Methyl 2-chloro-5-(methylamino)benzoate
Formel: C₉H₁₀ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD17291841
CAS-Nummer: 1340437-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR