Methyl 2-iodo-3-methylbenzoate 98%

Lieferant: Apollo Scientific
Achtung

OR55476-5G
APOSOR55476-5GEA 144 EUR
APOSOR55476-5G
Methyl 2-iodo-3-methylbenzoate 98%
Methyl 2-iodo-3-methylbenzoate
Formel: C₉H₉IO₂
Molecular Weight: 276,071 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12923249
CAS-Nummer: 103441-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR