1-Boc-6-methyl-2-indolboronsäurepinakolester 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methyl-1H-indole-2-boronic acid, N-BOC protected, (1-(tert-Butoxycarbonyl)-6-methyl-1H-indol-2-yl)boronic acid
OR5548-1G
APOSOR5548-1GEA
181
EUR
APOSOR5548-1G
1-Boc-6-methyl-2-indolboronsäurepinakolester 98%
1-Boc-6-methyl-2-indolboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₄ Molecular Weight: 275,11 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04973738 CAS-Nummer: 850568-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...