4-Methyl-3-(hydroxymethyl)morpholine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (4-Methylmorpholin-3-yl)methanol

OR55495-1G
APOSOR55495-1GEA 194 EUR
APOSOR55495-1G
4-Methyl-3-(hydroxymethyl)morpholine 95%
4-Methyl-3-(hydroxymethyl)morpholine
Formel: C₆H₁₃NO₂
Molecular Weight: 131,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26792530
CAS-Nummer: 1159598-86-3
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR