(S)-4-Methyl-3-(hydroxymethyl)morpholine 95%

Lieferant: Apollo Scientific

Synonyme: (S)-(4-Methylmorpholin-3-yl)methanol

OR55496-1G
APOSOR55496-1GEA 235 EUR
APOSOR55496-1G
(S)-4-Methyl-3-(hydroxymethyl)morpholine 95%
(S)-4-Methyl-3-(hydroxymethyl)morpholine
Formel: C₆H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD26792482
CAS-Nummer: 1620510-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR